Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RMND1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP156872
Description
RMND1 Polyclonal specifically detects RMND1 in Human samples. It is validated for Western Blot.Specifications
| RMND1 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 2% Sucrose with 0.09% Sodium Azide | |
| bA351K16, bA351K16.3, C6orf96, chromosome 6 open reading frame 96, FLJ20627, MGC117362, MGC149570, MGC88260, required for meiotic nuclear division 1 homolog (S. cerevisiae), required for meiotic nuclear division protein 1 homolog, RMD1 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 55005 | |
| Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| 0.5 mg/ml | |
| Western Blot 1.0 ug/ml | |
| Q9NWS8 | |
| RMND1 | |
| Synthetic peptides corresponding to RMND1(required for meiotic nuclear division 1 homolog (S. cerevisiae)) The peptide sequence was selected from the middle region of RMND1. Peptide sequence LEKHEIQPYEIALVHWENEELNYIKIEGQSKLHRGEIKLNSELDLDDAIL The peptide sequence for this immunogen was taken from within the described region. | |
| 100 μL | |
| Primary | |
| Expected identity based on immunogen sequence: Human: 100%; Mouse: 100%; Rat: 100%; Bovine: 92%; Equine: 92%; Rabbit: 92%; Chicken: 85%; Canine: 85%. | |
| Human, Mouse, Rat, Bovine, Canine, Equine, Guinea Pig, Rabbit | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction