Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RMND1 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$487.50
Specifications
| Antigen | RMND1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
RMND1 Polyclonal specifically detects RMND1 in Human samples. It is validated for Western Blot.Specifications
| RMND1 | |
| Polyclonal | |
| Rabbit | |
| Q9NWS8 | |
| 55005 | |
| IgG |
| Western Blot | |
| Unconjugated | |
| RUO | |
| bA351K16, bA351K16.3, C6orf96, chromosome 6 open reading frame 96, FLJ20627, MGC117362, MGC149570, MGC88260, required for meiotic nuclear division 1 homolog (S. cerevisiae), required for meiotic nuclear division protein 1 homolog, RMD1 | |
| RMND1 | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title