Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RMND5A Antibody (OTI2C8), DyLight 650, Novus Biologicals™
SDP

Catalog No. NBP273918C Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP273918C 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP273918C Supplier Novus Biologicals Supplier No. NBP273918C
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RMND5A Monoclonal specifically detects RMND5A in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen RMND5A
Applications Western Blot
Classification Monoclonal
Clone OTI2C8
Conjugate DyLight 650
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias C-terminal to LisH motif, 44 kDa, CTLH, FLJ12753, FLJ13910, FLJ21795, MGC78451, p44CTLH, protein RMD5 homolog A, required for meiotic nuclear division 5 homolog A (S. cerevisiae), RMD5
Gene Symbols RMND5A
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 1-249 of human RMND5A (NP_073617) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 64795.0
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.