Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RMND5A Monoclonal Antibody (OTI3D1), TrueMAB™, OriGene
SDP

Catalog No. 501679600
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-9600 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-9600 Supplier OriGene Supplier No. TA803152
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RMND5A is a protein coding gene.

Specifications

Antigen RMND5A
Applications Western Blot
Classification Monoclonal
Clone OTI3D1
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RMND5A
Gene Accession No. Q9H871
Gene Alias CTLH, GID2, GID2A, RMD5, p44CTLH
Gene Symbols RMND5A
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-249 of human RMND5A produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64795
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.