Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RN 1 dihydrochloride
GREENER_CHOICE

Catalog No. p-4835959
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

LSD1 inhibitor

Potent lysine specific demethylase 1 (LSD1) inhibitor (IC50 = 70 nM). Exhibits selectivity for LSD1 over MAO-A and MAO-B (IC50 values are 0.51 and 2.79 μM for MAO-A and MAO-B, respectively). Impairs long-term, but not short-term memory in mice. Brain penetrant following systemic administration.

Chemical Identifiers

Molecular Formula C23H29N3O2·2HCl

Specifications

Quantity 10 mg
Formula Weight 452.42
Chemical Name or Material RN 1 dihydrochloride
Recommended Storage Store at -20°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.