Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Tocris Bioscience™ RN 1734

Click to view available options
Quantity:
5 mg
25 mg
Description
RN 1734 is a selective TRPV4 antagonist (IC50 values are 2.3, 3.2 and 5.9 μM for hTRPV4, rTRPV4 and mTRPV4 receptors respectively). Displays selectivity for TRPV4 over other TRP channels (IC50 values are 2.3, >30, >30 and >100 μM for TRPV4, TRPV3, TRPM8 and TRPV1 respectively). Blocks TRPV4-mediated arteriole vasodilation and increases myogenic tone ex vivo.
Specifications
Specifications
| Chemical Name or Material | 2,4-Dichloro-N-isopropyl-N-(2-isopropylaminoethyl)benzenesulfonamide |
| CAS | 946387-07-1 |
| Quantity | 25 mg |
| Molecular Formula | C14H22Cl2N2O2S |
| Formula Weight | Observed MW: 353.31 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction