Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ RN 9893 hydrochloride
GREENER_CHOICE

Catalog No. p-7110873
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

Potent and selective TRPV4 antagonist

RN 9893 hydrochloride is a potent and selective TRPV4 antagonist (IC50 values are 320, 420 and 660 nM for mouse, human and rat channels, respectively). Exhibits >15-fold selectivity for TRPV4 over TRPV1, TRPV3 and TRPM8, and a panel of 54 other common biological targets. Orally bioavailable.

Specifications

Chemical Name or Material N-[4-[[4-(1-Methylethyl)-1-piperazinyl]sulfonyl]phenyl]-2-nitro-4-(trifluoromethyl)benzamide hydrochloride
CAS 2109450-40-8
Quantity 10 mg
Target TRPV Antagonists
Molecular Formula C21H23F3N4O5S.HCl
Formula Weight 536.95
Purity >99%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.