Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNA Control 250 with Manual

Catalog No. p-7180379
Encompass_Preferred
Click to view available options
Quantity:
2 x 20 μL, with Manual
2 x 20μL

Includes

RNA Control and THE RNA Storage Solution

The Invitrogen RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers.

The Invitrogen RNA Control 250 provides a precise, accurate, and reproducible RNA concentration reference for use in conjunction with NanoDrop™ Spectrophotometers. It is provided in two tubes containing 20 μL each. RNA Control 250 consists of a single, ultra-pure, 2000nt RNA transcript of a precisely defined sequence, molecular weight, and concentration. It is packaged in special non-stick tubes and includes the Invitrogen THE RNA Storage Solution. The RNA Control 250 will maintain ± 5% of stated concentration and >85% RNA integrity through 5 freeze/thaw cycles (10 duplicate measurements).

• A stable, highly pure transcript at a precisely known concentration
• Used to verify the accuracy of NanoDrop Spectrophotometer measurements
• Provided with THE RNA Storage Solution for blank measurements
• Enough for 20 measurements

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY

Specifications

Content And Storage RNA Control and THE RNA Storage Solution should be stored at –20°C
Format Tube
Label or Dye Unconjugated
Product Line Ambion™
Quantity 2 x 20μL
Shipping Condition Dry Ice
Volume 2 x 20 μL

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.