Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA Helicase A Antibody, Novus Biologicals™
SDP

Catalog No. NB176662 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB176662 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB176662 Supplier Novus Biologicals Supplier No. NBP312975
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

RNA Helicase A Polyclonal antibody specifically detects RNA Helicase A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)

Specifications

Antigen RNA Helicase A
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS, 1% BSA, 20% Glycerol (pH7)
Gene Alias DDX9ATP-dependent RNA helicase A, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A, nuclear DNAhelicase II; leukophysin), DEAH (Asp-Glu-Ala-His) box polypeptide 9, DEAH box protein 9, EC 3.6.1, EC 3.6.1.15, EC 3.6.4.13, FLJ17406, leukophysin, LKP, NDH II, NDH2, NDHII, Nuclear DNA helicase II, RHA
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the C-terminus region of human RNA Helicase A. The exact sequence is proprietary.
Purification Method Antigen Affinity-purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Core ESC Like Genes, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 1660
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.