Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNA Helicase A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$382.00
Specifications
Antigen | RNA Helicase A |
---|---|
Dilution | Western Blot 0.04-0.4 μg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Description
RNA Helicase A Polyclonal antibody specifically detects RNA Helicase A in Human samples. It is validated for Western BlotSpecifications
RNA Helicase A | |
Western Blot | |
Unconjugated | |
Rabbit | |
Core ESC Like Genes, Stem Cell Markers | |
PBS (pH 7.2) and 40% Glycerol | |
1660 | |
IgG | |
Affinity purified |
Western Blot 0.04-0.4 μg/mL | |
Polyclonal | |
Purified | |
RUO | |
Human | |
DDX9ATP-dependent RNA helicase A, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9, DEAD/H (Asp-Glu-Ala-Asp/His) box polypeptide 9 (RNA helicase A, nuclear DNAhelicase II; leukophysin), DEAH (Asp-Glu-Ala-His) box polypeptide 9, DEAH box protein 9, EC 3.6.1, EC 3.6.1.15, EC 3.6.4.13, FLJ17406, leukophysin, LKP, NDH II, NDH2, NDHII, Nuclear DNA helicase II, RHA | |
This antibody was developed against a recombinant protein corresponding to the following amino acid sequence: TIYIKQLGRRIFAREHGSNKKLAAQSCALSLVRQLYHLGVVEAYSGLTKKKEGETVEPYKVNLSQDLEHQLQNIIQELNLEILPPPEDPSVPVALNIGKLAQFEPSQ | |
Primary | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title