Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA Polymerase II/POLR2A Antibody (4H8), PE, Novus Biologicals™
SDP

Catalog No. NB200598PE Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB200598PE 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB200598PE Supplier Novus Biologicals Supplier No. NB200598PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNA Polymerase II/POLR2A Monoclonal specifically detects RNA Polymerase II/POLR2A in Human, Mouse, Yeast, Primate (Negative) samples. It is validated for Flow Cytometry, Flow (Intracellular).

Specifications

Antigen RNA Polymerase II/POLR2A
Applications Flow Cytometry
Classification Monoclonal
Clone 4H8
Conjugate PE
Dilution Flow Cytometry, Flow (Intracellular)
Gene Accession No. P24928
Gene Alias DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2
Gene Symbols POLR2A
Host Species Mouse
Immunogen 10 repeats of synthetic peptide YSPTSPS using chemically synthesized phospho-Ser 5 YSPTSpPS (Human). [UniProt# P24928]
Molecular Weight of Antigen 217 kDa
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, microRNA, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5430
Test Specificity Please note that this antibody detects both unphosphorylated and phosphorylated forms of the protein. When using this antibody in western blot and IP, it recognizes both the II0 and IIA form of RPB1 (ie the largest RNAPII subunit). These different forms of RPB1 (as seen in gels) have traditionally been loosely defined as 'hyper-' and 'hypo-phosphorylated', respectively. Here, however, it is important to remember that RPB1 contains no less than 52 repeats of a heptapeptide (TSPTSPS) that can be phosphorylated at different positions (Tyr1, Ser2, Thr4, Ser5 and Ser7; this antibody was raised against a synthetic peptide with 10 of these repeats, mostly but probably not entirely, chemically phosphorylated at Ser5). So, when we say that it recognizes both forms (II0/hyper-phos and IIA/hypo-phos), it might really just mean that the antibody recognizes a few phosphorylated Ser5s among 52 otherwise mostly un-phosphorylated repeats. Alternatively, it might conceivably (though less likely) recognize UN-phosphorylated repeats as there might conceivably be a few of those even in the II0/hyper-phos RPB1 form.'
Target Species Human, Mouse, Yeast, Primate (Negative)
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.