Learn More
Description
Specifications
Specifications
| Antigen | RNA Polymerase II/POLR2A |
| Applications | Western Blot, ChIP Assay, KnockDown |
| Classification | Monoclonal |
| Clone | C.15100055 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:500, Chromatin Immunoprecipitation (ChIP) 2 uL/IP, Chromatin Immunoprecipitation Sequencing 2 uL/IP, Knockdown Validated |
| Formulation | Ascites Fluid with 0.05% Sodium Azide |
| Gene Alias | DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2 |
| Gene Symbols | POLR2A |
| Host Species | Mouse |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
