Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA Polymerase II/POLR2A Antibody (C.15200004), Novus Biologicals™
SDP

Catalog No. NBP259213 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP259213 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP259213 Supplier Novus Biologicals Supplier No. NBP259213
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNA Polymerase II/POLR2A Monoclonal antibody specifically detects RNA Polymerase II/POLR2A in Human, Xenopus, Yeast, Zebrafish samples. It is validated for Western Blot, ChIP assay, ELISA, Immunocytochemistry, Immunofluorescence, Knockdown.

Specifications

Antigen RNA Polymerase II/POLR2A
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence, ChIP Assay
Classification Monoclonal
Clone C.15200004
Conjugate Unconjugated
Dilution Western Blot 1:1000, ELISA 1:3000, Immunocytochemistry/Immunofluorescence 1:500, Chromatin Immunoprecipitation (ChIP) 1 ug/IP, Chromatin Immunoprecipitation Sequencing 1ug/ChIP, Knockdown Validated
Formulation PBS with 0.05% Sodium Azide
Gene Alias DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2
Gene Symbols POLR2A
Host Species Mouse
Immunogen The exact immunogen is propietary information.
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, microRNA, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5430
Target Species Human, Xenopus species, Yeast, Zebrafish
Content And Storage Store at -20C short term. Aliquot and store at -80C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.