Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA Polymerase II/POLR2A Antibody, Novus Biologicals™
SDP

Catalog No. NBP232080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP232080 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP232080 Supplier Novus Biologicals Supplier No. NBP232080
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 3 publications

RNA Polymerase II/POLR2A Polyclonal specifically detects RNA Polymerase II/POLR2A in Human samples. It is validated for Chromatin Immunoprecipitation, Immunoprecipitation, Western Blot (Negative).

Specifications

Antigen RNA Polymerase II/POLR2A
Applications Immunoprecipitation, Western Blot, ChIP Assay
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 10 ug/mg lysate, Western Blot (Negative), Chromatin Immunoprecipitation (ChIP) 8 ug
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_000928.1
Gene Alias DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2
Gene Symbols POLR2A
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 754 to 804 of human RNA Polymerase II using the numbering given in entry NP_000928.1 (GeneID 5430).
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, microRNA, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5430
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with X. tropicalis, Panda, Orangutan, Rhesus Monkey, Gorilla, Chimpanzee, Cetartiodactyla, Eukaryota, Green anole and Southern platyfish.
Target Species Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.