Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA Polymerase II/POLR2A Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB290103 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290103 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290103 Supplier Novus Biologicals Supplier No. NBP232080PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNA Polymerase II/POLR2A Polyclonal antibody specifically detects RNA Polymerase II/POLR2A in Human samples. It is validated for Immunoprecipitation, Western Blot (Negative), ChIP assay (ChIP)

Specifications

Antigen RNA Polymerase II/POLR2A
Applications Immunoprecipitation, ChIP Assay
Classification Polyclonal
Conjugate PerCP
Dilution Immunoprecipitation, Western Blot (Negative), Chromatin Immunoprecipitation (ChIP)
Formulation PBS
Gene Alias DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2
Host Species Rabbit
Immunogen The immunogen this antibody was made to, maps to a region between residue 754 to 804 of human RNA Polymerase II using the numbering given in entry NP_000928.1 (GeneID 5430).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5430
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.