Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA Polymerase II/POLR2A, p Ser2 Antibody, Novus Biologicals™
SDP

Catalog No. NB1001805 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1001805 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1001805 Supplier Novus Biologicals Supplier No. NB1001805
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 13 publications

RNA Polymerase II/POLR2A (p Ser2) Polyclonal specifically detects RNA Polymerase II/POLR2A (p Ser2) in Human, Mouse, Yeast samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Single Cell Western, Western Blot.

Specifications

Antigen RNA Polymerase II/POLR2A (p Ser2)
Applications Immunohistochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Proximity Ligation Assay, Single Cell Western, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunohistochemistry 1:200 - 1:1000, Immunoprecipitation 2-10 mg/mg, Immunohistochemistry-Paraffin 1:200 - 1:1000, Single Cell Western 100 ug/mL
Formulation TBS and 0.1% BSA with 0.09% Sodium Azide
Gene Accession No. P24928
Gene Alias DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2
Gene Symbols POLR2A
Host Species Rabbit
Immunogen Immunogen for this antibody was a phosphorylated synthetic peptide, which represented a portion of human RNA Polymerase II (GeneID 5430) around serine 2 of the C-terminal repeat YSPTSPS.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, microRNA, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5430
Test Specificity RNA Polymerase 2 Phosphospecific [Ser2]
Target Species Human, Mouse, Yeast
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.