Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNA Polymerase II/POLR2A, p Thr4 Antibody, Alexa Fluor™ 405, Novus Biologicals™
SDP

Catalog No. NBP149546W Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP149546W 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP149546W Supplier Novus Biologicals Supplier No. NBP149546AF405
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNA Polymerase II/POLR2A Polyclonal specifically detects RNA Polymerase II/POLR2A in Human, Chicken, Drosophila, Yeast samples. It is validated for Western Blot.

Specifications

Antigen RNA Polymerase II/POLR2A (p Thr4)
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 405
Dilution Western Blot
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Accession No. P24928
Gene Alias DNA-directed RNA polymerase II largest subunit, RNA polymerase II 220 kdsubunit, DNA-directed RNA polymerase II subunit A, DNA-directed RNA polymerase II subunit RPB1, DNA-directed RNA polymerase III largest subunit, EC 2.7.7.48, EC 2.7.7.6, hRPB220, hsRPB1, MGC75453, POLR2, POLRA, polymerase (RNA) II (DNA directed) polypeptide A (220kD), polymerase (RNA) II (DNA directed) polypeptide A, 220kDa, RNA Pol 2, RNA Polymerase 2, RNA polymerase II subunit B1, RNA-directed RNA polymerase II subunit RPB1, RNAPII, RPB1, RPBh1, RpIILS, RPO2, RPOL2
Gene Symbols POLR2A
Host Species Rabbit
Immunogen A synthetic peptide made to a phosphorylated Threonine (position 4) of the CTD heptad in the human RNA polymerase II protein [UniProt P24928]
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Epigenetics, microRNA, Phospho Specific, Transcription Factors and Regulators
Primary or Secondary Primary
Gene ID (Entrez) 5430
Test Specificity This antibody is specific for the phosphorylated Threonine found in position 4 of the RNA polymerase II CTD heptad.
Target Species Human, Chicken, Drosophila, Yeast
Content And Storage Store at 4C in the dark.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.