Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNase A Solution, 10 mg/mL, Spectrum™ Chemical
SDP

Catalog No. 18606099 Shop All Spectrum Chemical Manufacturing Corporation Products
Click to view available options
Quantity:
1 mL
Packaging:
Amber Glass Bottle

R3032, 9001-99-4

Spectrum™ Chemical RNase A Solution, 10 mg/ml

Chemical Identifiers

CAS 9001-99-4
MDL Number MFCD00132181
IUPAC Name Ribonuclease A

Specifications

CAS Max % 1
Packaging Amber Glass Bottle
Quantity 1 mL
Grade Reagent
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.