Learn More
RNase L Mouse anti-Human, Unconjugated, Clone: 2E9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier:  MilliporeSigma MABS2055100UG
 View more versions of this product
Catalog No. MABS20551MI
Only null left
Add to Cart
Description
Description
RNase L Monoclonal antibody specifically detects RNase L in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blotting.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
RNase L
Monoclonal
Unconjugated
2-5A-dependent ribonuclease;2-5A-dependent RNase;Ribonuclease 4;Ribonuclease L
Mouse
∼83.53 kDa calculated.
100 μg
Primary
Human
Purified
ELISA, Immunohistochemistry (Paraffin), Western Blot
2E9
NP_066956
RNASEL, RNS4
Full length recombinant human 2-5A-dependent ribonuclease (RNase L).
Protein G Purified
Signaling
Clone 2E9 is a mouse monoclonal antibody that detects Ribonuclease L in human cells.
Stable for 1 year at 2 to 8°C from date of receipt.
IgG1 κ
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Certifications
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.