Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNase L Mouse anti-Human, Unconjugated, Clone: 2E9, MilliporeSigma™
Mouse Monoclonal Antibody
$189.29 - $477.80
Specifications
Antigen | RNase L |
---|---|
Clone | 2E9 |
Applications | ELISA, Immunohistochemistry (Paraffin), Western Blot |
Classification | Monoclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MABS20552MI
|
MilliporeSigma
MABS205525UG |
25 μg |
Each for $189.29
|
|
|||||
MABS20551MI
|
MilliporeSigma
MABS2055100UG |
100 μg |
Each for $477.80
|
|
|||||
Description
RNase L Monoclonal antibody specifically detects RNase L in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blotting.Specifications
RNase L | |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
Unconjugated | |
Mouse | |
Human | |
2-5A-dependent ribonuclease;2-5A-dependent RNase;Ribonuclease 4;Ribonuclease L | |
Full length recombinant human 2-5A-dependent ribonuclease (RNase L). | |
Primary | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
∼83.53 kDa calculated. |
2E9 | |
Monoclonal | |
Purified | |
Signaling | |
NP_066956 | |
RNASEL, RNS4 | |
IgG1 κ | |
Protein G Purified | |
Clone 2E9 is a mouse monoclonal antibody that detects Ribonuclease L in human cells. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title