Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNase L Mouse anti-Human, Unconjugated, Clone: 2E9, MilliporeSigma™
Mouse Monoclonal Antibody
Supplier: MilliporeSigma MABS205525UG
Description
RNase L Monoclonal antibody specifically detects RNase L in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin), Western Blotting.
Specifications
RNase L | |
Monoclonal | |
Unconjugated | |
2-5A-dependent ribonuclease;2-5A-dependent RNase;Ribonuclease 4;Ribonuclease L | |
Mouse | |
∼83.53 kDa calculated. | |
25 μg | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin), Western Blot | |
2E9 | |
NP_066956 | |
RNASEL, RNS4 | |
Full length recombinant human 2-5A-dependent ribonuclease (RNase L). | |
Protein G Purified | |
Signaling | |
Clone 2E9 is a mouse monoclonal antibody that detects Ribonuclease L in human cells. | |
Stable for 1 year at 2 to 8°C from date of receipt. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction