Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ RNase T1 (1000 U/μL)

Catalog No. FEREN0542
Click to view available options
Quantity:
100,000 units
500,000 units

RNase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues.

Thermo Scientific Rnase T1 is an endoribonuclease that specifically degrades single-stranded RNA at G residues (see Figure 1 in Supporting Data). It cleaves the phosphodiester bond between the 3'-guanylic residue and the 5'-OH residue of adjacent nucleotides with the formation of corresponding intermediate 2', 3'-cyclic phosphate. The reaction products are 3'-GMP and oligonucleotides with a terminal 3'-GMP.

Rnase T1 does not require metal ions for activity.

Applications

  • Removal of RNA from DNA preparations
  • RNA sequencing
  • Ribonuclease protection assays. Used in conjunction with RNase A
  • Removal of RNA from recombinant protein preparations
  • Determination of the level of RNA transcripts synthesized in vitro from DNA templates containing a "G-less cassette"
TRUSTED_SUSTAINABILITY

Specifications

Concentration 1000 U/μL
Enzyme RNase
Quantity 500,000 units
Product Type RNase T1
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.