Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RNASE7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5121638
This item is not returnable.
View return policy
Description
RNASE7 Polyclonal antibody specifically detects RNASE7 in Human, Rat samples. It is validated for Western Blot.
Specifications
RNASE7 | |
Polyclonal | |
Unconjugated | |
RNASE7 | |
RAE1; Ribonuclease 7; ribonuclease A family member 7; ribonuclease, RNase A family, 7; RNase 7; RNASE7; SAP-2; skin-derived antimicrobial protein 2; UNQ2516/PRO6006 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
84659 | |
-20°C | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
Q9H1E1 | |
RNASE7 | |
Lys31-Val155 (Accession Q9H1E1), with N-terminal His Tag. | |
100 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction