Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNASE7 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
$209.50 - $511.50
Specifications
Antigen | RNASE7 |
---|---|
Concentration | 0.5 mg/mL |
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
PIPA5121639
|
Invitrogen™
PA5121639 |
20 μL |
Each for $209.50
|
|
|||||
PIPA5121638
|
Invitrogen™
PA5121638 |
100 μL |
Each for $511.50
|
|
|||||
Description
RNASE7 Polyclonal antibody specifically detects RNASE7 in Human, Rat samples. It is validated for Western Blot.Specifications
RNASE7 | |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Rat | |
Q9H1E1 | |
84659 | |
Lys31-Val155 (Accession Q9H1E1), with N-terminal His Tag. | |
Primary | |
-20°C | |
RNASE7 |
0.5 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.05% ProClin 300 | |
RAE1; Ribonuclease 7; ribonuclease A family member 7; ribonuclease, RNase A family, 7; RNase 7; RNASE7; SAP-2; skin-derived antimicrobial protein 2; UNQ2516/PRO6006 | |
RNASE7 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title