Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNASE7 Polyclonal Antibody, Invitrogen™

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5121639

 View more versions of this product

Catalog No. PIPA5121639


Only null left
Add to Cart
This item is not returnable. View return policy

Description

Description

RNASE7 Polyclonal antibody specifically detects RNASE7 in Human, Rat samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

RNASE7
Polyclonal
Unconjugated
RNASE7
RAE1; Ribonuclease 7; ribonuclease A family member 7; ribonuclease, RNase A family, 7; RNase 7; RNASE7; SAP-2; skin-derived antimicrobial protein 2; UNQ2516/PRO6006
Rabbit
Antigen affinity chromatography
RUO
84659
-20°C
Liquid
Western Blot
0.5 mg/mL
PBS with 50% glycerol and 0.05% ProClin 300
Q9H1E1
RNASE7
Lys31-Val155 (Accession Q9H1E1), with N-terminal His Tag.
20 μL
Primary
Human, Rat
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.