Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNASEH2A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$492.89
Specifications
| Antigen | RNASEH2A |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP158235
![]() |
Novus Biologicals
NBP158235 |
100 μL |
Each for $492.89
|
|
|||||
NBP15823520
![]() |
Novus Biologicals
NBP15823520UL |
20 μL | N/A | N/A | N/A | ||||
Description
RNASEH2A Polyclonal specifically detects RNASEH2A in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| RNASEH2A | |
| Polyclonal | |
| Purified | |
| RUO | |
| AGS4RNase H2 subunit A, Aicardi-Goutieres syndrome 4, Aicardi-Goutieres syndrome 4 protein, JUNB, ribonuclease H2 subunit A, ribonuclease H2, large subunit, ribonuclease H2, subunit A, Ribonuclease HI large subunit, Ribonuclease HI subunit A, ribonuclease HI, large subunit, RNase H(35), RNASEHIEC 3.1.26.4, RNHIARNase HI large subunit, RNHL | |
| RNASEH2A | |
| IgG | |
| Protein A purified |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| O75792 | |
| 10535 | |
| Synthetic peptides corresponding to RNASEH2A (ribonuclease H2, subunit A) The peptide sequence was selected from the middle region of RNASEH2A. Peptide sequence AQTILEKEAEDVIWEDSASENQEGLRKITSYFLNEGSQARPRSSHRYFLE. | |
| Primary | |
| This product is specific to Subunit or Isoform: A. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title