Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RND3 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB318707 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB318707 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB318707 Supplier Novus Biologicals Supplier No. NBP298843APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RND3 Polyclonal antibody specifically detects RND3 in Human samples. It is validated for Western Blot

Specifications

Antigen RND3
Applications Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot
Formulation PBS
Gene Alias ARHE, Protein MemB, Rho family GTPase 3member E, RHO8, RHOE, Rho-related GTP-binding protein Rho8, rho-related GTP-binding protein RhoE, Rnd3, small GTP binding protein Rho8
Host Species Rabbit
Immunogen Produced in rabbits immunized with a synthetic peptide corresponding to the C-terminus of the Human RND3.
Purification Method Antigen and protein A Affinity-purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 390
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.