Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF113A Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | RNF113A |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP18031820
![]() |
Novus Biologicals
NBP18031820UL |
20 μL |
Each for $206.00
|
|
|||||
NBP180318
![]() |
Novus Biologicals
NBP180318 |
100 μL |
Each for $487.50
|
|
|||||
Description
RNF113A Polyclonal specifically detects RNF113A in Human samples. It is validated for Western Blot.Specifications
RNF113A | |
Polyclonal | |
Rabbit | |
Zinc Finger | |
ring finger protein 113A, RNF113Cwc24, Zinc finger protein 183, zinc finger protein 183 (RING finger, C3HC4 type), ZNF183 | |
RNF113A | |
IgG |
Western Blot | |
Unconjugated | |
RUO | |
NP_008909 | |
7737 | |
Synthetic peptide directed towards the middle region of human RNF113A. Peptide sequence LQHFRTTPRCYVCDQQTNGVFNPAKELIAKLEKHRATGEGGASDLPEDPD. | |
Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title