Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF133 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NBP162487

 View more versions of this product

Catalog No. NBP162487


Only null left
Add to Cart

Description

Description

RNF133 Polyclonal specifically detects RNF133 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RNF133
Polyclonal
Unconjugated
PBS, 2% Sucrose with 0.09% Sodium Azide
E3 ubiquitin-protein ligase RNF133, EC 6.3.2.-, ring finger protein 133MGC27072
Rabbit
42 kDa
100 μL
Zinc Finger
168433
Human, Mouse, Rat, Equine, Rabbit
IgG
Western Blot
0.5 mg/ml
Western Blot 1.0 ug/ml
Q8WVZ7
RNF133
Synthetic peptides corresponding to RNF133(ring finger protein 133) The peptide sequence was selected from the N terminal of RNF133. Peptide sequence VVWMAYMNISFHVGNHVLSELGETGVFGRSSTLKRVAGVIVPPEGKIQNA.
Affinity purified
RUO
Primary
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 50μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer.
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.