Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF146 Antibody, Novus Biologicals™
SDP

Catalog No. NBP155068 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP155068 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP155068 Supplier Novus Biologicals Supplier No. NBP155068
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF146 Polyclonal specifically detects RNF146 in Human samples. It is validated for Western Blot.

Specifications

Antigen RNF146
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Dilution Western Blot 1.0 ug/ml
Formulation PBS, 2% Sucrose with 0.09% Sodium Azide
Gene Accession No. Q9NTX7
Gene Alias dactylidin, dJ351K20.1, DKFZp434O1427, ring finger protein 146,2610509H23Rik, RP3-351K20.1
Gene Symbols RNF146
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RNF146(ring finger protein 146) The peptide sequence was selected from the C terminal of RNF146 (NP_112225). Peptide sequence AVVAQHSLTQQRLLVSNANQTVPDRSDRSGTDRSVAGGGTVSVSVRSRRP.
Molecular Weight of Antigen 39 kDa
Purification Method Protein A purified
Quantity 100 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 81847
Reconstitution Reconstitute in 100μL distilled water to a final antibody concentration of 1mg/mL.
Target Species Human, Mouse, Rat, Canine, Equine, Guinea Pig
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.