Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF146 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578565
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA578565 100 μL
1 options

Catalog No. PIPA578565

Supplier: Invitrogen™ PA578565

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: mouse brain, rat brain Predicted Reactivity: Human (91%), Rhesus Monkey (91%), Bovine (87%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RNF146 protein function is currently unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF146
Applications Immunohistochemistry (Frozen), Western Blot
Classification Polyclonal
Concentration 0.79 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene RNF146
Gene Accession No. Q5XIK5, Q9CZW6
Gene Alias 2610509H23Rik; dactylidin; E3 ubiquitin-protein ligase RNF146; iduna; RING finger protein 146; RING finger protein 146; E3 ubiquitin-protein ligase RNF146; RING-type E3 ubiquitin transferase RNF146; Rnf146
Gene Symbols RNF146
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of mouse RNF146. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 308051, 68031
Target Species Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.