Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RNF152 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA595817
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
PIPA595817 100 μL
1 options

Catalog No. PIPA595817

Supplier: Invitrogen™ PA595817

Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: YMLPLPISKE RALLPGDMGC RLLPGSQQKS VTVVTIPAEQ QPLQGGAPQE AVEEEQDRRG VVKSSTWSGV CTVILVACVL VFLLGIVLHN MSCISKRFTV I; Positive Samples: U-251MG, 293T, LO2; Cellular Location: Lysosome membrane, Single-pass membrane protein.

TRUSTED_SUSTAINABILITY

Specifications

Antigen RNF152
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 1.61 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3
Gene RNF152
Gene Accession No. Q8N8N0
Gene Alias E3 ubiquitin-protein ligase RNF152; RING finger protein 152; RING-type E3 ubiquitin transferase RNF152; RNF152
Gene Symbols RNF152
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 100-200 of human RNF152 (NP_7758281).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 220441
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.