Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF175 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP155079
Description
RNF175 Polyclonal specifically detects RNF175 in Human samples. It is validated for Western Blot.Specifications
RNF175 | |
Polyclonal | |
Unconjugated | |
PBS, 2% Sucrose with 0.09% Sodium Azide | |
FLJ34190, ring finger protein 175 | |
Rabbit | |
19 kDa | |
100 μL | |
Zinc Finger | |
285533 | |
Human | |
Purified |
Western Blot | |
1 mg/ml | |
Western Blot 1.0 ug/ml | |
Q8NB61 | |
RNF175 | |
Synthetic peptides corresponding to RNF175(ring finger protein 175) The peptide sequence was selected from the middle region of RNF175. Peptide sequence YGLYYGVMGRDFAEICSDYMASTIGFYSVSRLPTRSLSDNICAVCGQKII. | |
Protein A purified | |
RUO | |
Primary | |
Centrifuge the vial of lyoph antibody at 12,000 x g for 20 seconds. Add 100μL of distilled water. Vortex followed by centrifuge again to pellet the solution.Final concentration is 1mg/mL in PBS buffer. | |
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction