Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF182 Antibody, Novus Biologicals™
SDP

Catalog No. NBP15976220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP15976220 20 μL
NBP159762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP15976220 Supplier Novus Biologicals Supplier No. NBP15976220UL

Rabbit Polyclonal Antibody

RNF182 Polyclonal specifically detects RNF182 in Human samples. It is validated for Western Blot.

Specifications

Antigen RNF182
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q8N6D2
Gene Alias E3 ubiquitin-protein ligase RNF182, EC 6.3.2.-, FLJ40772, ring finger protein 182MGC33993
Gene Symbols RNF182
Host Species Rabbit
Immunogen Synthetic peptides corresponding to RNF182(ring finger protein 182) The peptide sequence was selected from the N terminal of RNF182. Peptide sequence CLPDDEVSSLPDDNNILVNLTCGGKGKKCLPENPTELLLTPKRLASLVSP.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 221687
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.