Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF182 Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

$206.00 - $487.50

Specifications

Antigen RNF182
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Host Species Rabbit
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NBP15976320
SDP
View Documents
Novus Biologicals
NBP15976320UL
20 μL
Each for $206.00
Only null left
Add to Cart
 
NBP159763
SDP
View Documents
Novus Biologicals
NBP159763
100 μL
Each for $487.50
Only null left
Add to Cart
 
Description

Description

RNF182 Polyclonal specifically detects RNF182 in Human samples. It is validated for Western Blot.
Specifications

Specifications

RNF182
Polyclonal
Rabbit
Zinc Finger
E3 ubiquitin-protein ligase RNF182, EC 6.3.2.-, FLJ40772, ring finger protein 182MGC33993
RNF182
IgG
Western Blot
Unconjugated
RUO
Q8N6D2
221687
Synthetic peptides corresponding to RNF182(ring finger protein 182) The peptide sequence was selected from the middle region of RNF182. Peptide sequence LSSTPVVEFYRPASFDSVTTVSHNWTVWNCTSLLFQTSIRVLVWLLGLLY.
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.