Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF183 Antibody, Novus Biologicals™
SDP

Catalog No. NBP17419220 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP17419220 20 μL
NBP174192 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP17419220 Supplier Novus Biologicals Supplier No. NBP17419220UL

Rabbit Polyclonal Antibody

RNF183 Polyclonal specifically detects RNF183 in Mouse samples. It is validated for Western Blot.

Specifications

Antigen RNF183
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000
Formulation PBS & 2% Sucrose. with No Preservative
Gene Accession No. Q8QZS5
Gene Alias FLJ31197, MGC4734, ring finger protein 183
Gene Symbols RNF183
Host Species Rabbit
Immunogen Synthetic peptides corresponding to the N terminal of Rnf183. Immunizing peptide sequence LSLVTPARRRLLCPLCRQPTVLASGQPVTDLPTDTAMLTLLRLEPHHVIL.
Purification Method Affinity Purified
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 138065
Target Species Mouse
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.