Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF2 Antibody, DyLight 650, Novus Biologicals™
SDP

Catalog No. NB120062 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB120062 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB120062 Supplier Novus Biologicals Supplier No. NBP177826C
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RNF2 Polyclonal specifically detects RNF2 in Human, Mouse, Chinese Hamster samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RNF2
Applications Western Blot, ELISA, Immunofluorescence
Classification Polyclonal
Conjugate DyLight 650
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias BAP-1, BAP1RING finger protein 1B, DINGRING finger protein BAP-1, EC 6.3.2, HIPI3RING1BE3 ubiquitin-protein ligase RING2, Huntingtin-interacting protein 2-interacting protein 3, Protein DinG, ring finger protein 2HIP2-interacting protein 3, RING1b, RING2EC 6.3.2.-
Host Species Goat
Immunogen RNF2 Antibody was prepared from whole goat serum produced by repeated immunizations with a synthetic peptide corresponding to an internal region near aa 180-205 of human RNF2 protein. (Uniprot: Q99496)
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cancer, Signal Transduction, Transcription Factors and Regulators, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 6045
Target Species Human, Mouse, Chinese Hamster
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.