Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF20 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB292476 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB292476 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB292476 Supplier Novus Biologicals Supplier No. NB1002242APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF20 Polyclonal specifically detects RNF20 in Human, Mouse samples. It is validated for ChIP Assay, Immunoprecipitation, Western Blot.

Specifications

Antigen RNF20
Applications ChIP Assay, Immunoprecipitation, Western Blot
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation, Chromatin Immunoprecipitation (ChIP)
Formulation PBS
Gene Alias BRE1, BRE1 E3 ubiquitin ligase homolog, BRE1-A, BRE1AMGC129667, E3 ubiquitin-protein ligase BRE1A, EC 6.3.2, EC 6.3.2.-, FLJ11189, FLJ20382, hBRE1MGC129668, homolog of S. cerevisiae BRE1, KAIA2779, ring finger protein 20KIAA2779
Host Species Rabbit
Immunogen Maps to a region between residue 1 and 50 of human Ring Finger 20 using the numbering given in entry NP_062538.5 (GeneID 56254).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 56254
Target Species Human, Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.