Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RNF213 Rabbit anti-Human, Unconjugated, Polyclonal, MilliporeSigma™
Rabbit Polyclonal Antibody
$208.00 - $457.00
Specifications
| Antigen | RNF213 |
|---|---|
| Concentration | Please refer to lot specific datasheet. |
| Applications | Immunohistochemistry (Paraffin), Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|
ABN1474-25UG
|
MilliporeSigma
ABN147425UG |
25 μg |
Each for $208.00
|
|
|||||
|
ABN1474-100
|
MilliporeSigma
ABN1474100UG |
100 μg |
Each for $457.00
|
|
|||||
Description
Anti-RNF213, Cat. No. ABN1474, is a highly specific rabbit polyclonal antibody that targets E3 ubiquitin-protein ligase RNF213 and has been tested for use in Immunohistochemistry (Paraffin) and Western Blotting.Specifications
| RNF213 | |
| Immunohistochemistry (Paraffin), Western Blot | |
| Unconjugated | |
| RUO | |
| Human | |
| E3 ubiquitin-protein ligase RNF213;EC:2.3.2.27;ALK lymphoma oligomerization partner on chromosome 17;Mysterin;RING finger protein 213;RING-type E3 ubiquitin transferase RNF213 | |
| RNF213;ALO17;C17orf27;KIAA1554;KIAA1618;MYSTR | |
| IgG | |
| Affinity Purified | |
| This rabbit polyclonal antibody detects E3 ubiquitin-protein ligase RNF213 in human cells. It targets an epitope within 15 amino acids from the C-terminal half. | |
| ∽590 kDa observed; 591.41 kDa calculated. Uncharacterized bands may be observed in some lysate(s). |
| Please refer to lot specific datasheet. | |
| Polyclonal | |
| Rabbit | |
| Neuroscience | |
| Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide. | |
| NP_001243000 | |
| KLH-conjugated linear peptide corresponding to 15 amino acids from the C-terminal half of human E3 ubiquitin-protein ligase RNF213. Immunogen sequence is conserved in isoforms 1, 2, and 4. | |
| Primary | |
| Stable for 1 year at 2-8°C from date of receipt. | |
| Antibodies |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title