Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF23 Antibody (OTI2A6), Alexa Fluor™ 594, Novus Biologicals™
SDP

Catalog No. NP273926A94 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP273926A94 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP273926A94 Supplier Novus Biologicals Supplier No. NBP273926AF594
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNF23 Monoclonal specifically detects RNF23 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot.

Specifications

Antigen RNF23
Applications Western Blot
Classification Monoclonal
Clone OTI2A6
Conjugate Alexa Fluor 594
Dilution Western Blot
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias MGC32984, RING finger protein 23RNF23TFPtripartite motif-containing 39, Testis-abundant finger protein, TRIM39B, tripartite motif containing 39, tripartite motif-containing protein 39
Gene Symbols TRIM39
Host Species Mouse
Immunogen Recombinant protein fragment corresponding to amino acids 144-378 of human RNF23 (NP_742013) produced in E.coli.
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 56658.0
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.