Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF23 Antibody (OTI2A6), Novus Biologicals™
SDP

Catalog No. NBP246014 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP246014 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP246014 Supplier Novus Biologicals Supplier No. NBP246014
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Ensure accurate, reproducible results in Western Blot

RNF23 Monoclonal specifically detects RNF23 in Human, Mouse, Rat, Canine samples. It is validated for Western Blot.

Specifications

Antigen RNF23
Applications Western Blot
Classification Monoclonal
Clone OTI2A6
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000
Formulation PBS (pH 7.3), 1.0% BSA and 50% Glycerol with 0.02% Sodium Azide
Gene Alias MGC32984, RING finger protein 23RNF23TFPtripartite motif-containing 39, Testis-abundant finger protein, TRIM39B, tripartite motif containing 39, tripartite motif-containing protein 39
Gene Symbols TRIM39
Host Species Mouse
Immunogen Human recombinant protein fragment corresponding to amino acids 144-378 of human TRIM39 (NP_742013) produced in E.coli.
Molecular Weight of Antigen 56.2 kDa
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 56658
Target Species Human, Mouse, Rat, Canine
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.