Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF34 Antibody, Novus Biologicals™
SDP

Catalog No. NB394497 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB394497 25 μL
NBP256413 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB394497 Supplier Novus Biologicals Supplier No. NBP25641325UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody has been used in 2 publications

RNF34 Polyclonal specifically detects RNF34 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence, Immunoprecipitation.

Specifications

Antigen RNF34
Applications Western Blot, Immunocytochemistry, Immunofluorescence, Immunoprecipitation
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blot 0.04-0.4 μg/mL, Immunocytochemistry/Immunofluorescence 0.25-2 μg/mL, Immunoprecipitation
Formulation PBS, pH 7.2, containing 40% glycerol with 0.02% Sodium Azide
Gene Alias CARP1, Caspase regulator CARP1, Caspases-8 and -10-associated RING finger protein 1, E3 ubiquitin-protein ligase RNF34, EC 6.3.2, EC 6.3.2.-, FLJ21786, FYVE-RING finger protein Momo, hRFI, Human RING finger homologous to inhibitor of apoptosis protein, RFI, RIF, RIFF, ring finger protein 34°CARP-1, RING finger protein RIFF
Gene Symbols RNF34
Host Species Rabbit
Immunogen This antibody was developed against a recombinant protein corresponding to the following amino acid sequence:NIVCKACGLSFSVFRKKHVCCDCKKDFCSVCSVLQENLRRCSTCHLLQETAFQRPQLMRLKVKDL
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 80196
Test Specificity Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins.
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.