Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF34 Antibody, Novus Biologicals™
SDP

Catalog No. NBP177903 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.5 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP177903 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP177903 Supplier Novus Biologicals Supplier No. NBP177903
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF34 Polyclonal specifically detects RNF34 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen RNF34
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Polyclonal
Concentration LYOPH
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000, ELISA 1:4000-1:20000, Immunocytochemistry/Immunofluorescence 1:10-1:500
Formulation 0.02 M Potassium Phosphate, 0.15 M Sodium Chloride, pH 7.2 with 0.01% Sodium Azide
Gene Accession No. NP_919247
Gene Alias CARP1, Caspase regulator CARP1, Caspases-8 and -10-associated RING finger protein 1, E3 ubiquitin-protein ligase RNF34, EC 6.3.2, EC 6.3.2.-, FLJ21786, FYVE-RING finger protein Momo, hRFI, Human RING finger homologous to inhibitor of apoptosis protein, RFI, RIF, RIFF, ring finger protein 34CARP-1, RING finger protein RIFF
Gene Symbols RNF34
Host Species Rabbit
Immunogen This affinity purified RNF34 Antibody was prepared from whole rabbit serum produced by repeated immunizations with a recombinant protein corresponding to amino acids 1-373 of human RNF34 protein. (Uniprot: Q969K3)
Molecular Weight of Antigen 41.64 kDa
Purification Method Protein A purified
Quantity 0.5 mg
Regulatory Status RUO
Research Discipline Apoptosis
Primary or Secondary Primary
Gene ID (Entrez) 80196
Test Specificity This protein A purified antibody is directed against human RNF34 protein. A BLAST analysis was used to suggest cross-reactivity with RNF34 protein from orangutan (99%), bovine (91%), dog (89%), mouse (88%) and rat (85%) based on a high degree of protein:protein homology with the immunizing sequence. Expect reactivity with isoform 1 and 2 of RNF34. Reactivity against homologues from other sources is not known.
Reconstitution Reconstitute with 500 ul deionized water (or equivalent)
Target Species Human
Content And Storage Store lyophilized antibody at 4°C. Aliquot reconstituted liquid and store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.