Learn More
Description
Specifications
Specifications
| Antigen | RNF38 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100-1:2000 |
| Formulation | PBS and 2% Sucrose with 0.09% Sodium Azide |
| Gene Alias | FLJ21343, ring finger protein 38 |
| Gene Symbols | RNF38 |
| Host Species | Rabbit |
| Immunogen | Synthetic peptides corresponding to RNF38(ring finger protein 38) The peptide sequence was selected from the N terminal of RNF38. Peptide sequence FDYTSASPAPSPPMRPWEMTSNRQPPSVRPSQHHFSGERCNTPARNRRSP. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
