Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. p-7207457 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-100-2685 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-100-2685 Supplier Proteintech Group Inc Supplier No. 178101AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The protein encoded by this gene contains a RING finger motif and acts as a transcription regulator. This protein has been shown to interact with, and inhibit the activity of, TRPS1, a transcription suppressor of GATA-mediated transcription. Transcription repressor ZNF278/PATZ is found to interact with this protein, and thus reduce the enhancement of androgen receptor-dependent transcription mediated by this protein. Studies of the mouse and rat counterparts suggested a role of this protein in spermatogenesis. A pseudogene of this gene is found on chromosome 1.

Specifications

Antigen RNF4
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.32 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene RNF4
Gene Accession No. O88846, P78317, Q9QZS2
Gene Alias E3 ubiquitin ligase RNF4, Protein SNURF, RES4 26, ring finger protein 4, RNF4, SNURF
Gene Symbols RNF4
Host Species Rabbit
Immunogen RNF4 Fusion Protein Ag12115
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19822, 29274, 6047
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.