Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF40 Antibody, Novus Biologicals™
SDP

Catalog No. NBP220190 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP220190 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP220190 Supplier Novus Biologicals Supplier No. NBP220190
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF40 Polyclonal specifically detects RNF40 in Human samples. It is validated for Western Blot.

Specifications

Antigen RNF40
Applications Western Blot
Classification Polyclonal
Concentration 1mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:3000
Formulation PBS (pH 7.0), 1.0% BSA and 20% Glycerol with 0.01% Thimerosal
Gene Accession No. O75150
Gene Alias 95 kDa retinoblastoma-associated protein, BRE1-B, BRE1BMGC13051, EC 6.3.2,95 kDa retinoblastoma protein binding protein, EC 6.3.2.-, KIAA0661BRE1 E3 ubiquitin ligase homolog B, Rb-associated protein, RBP95E3 ubiquitin-protein ligase BRE1B, ring finger protein 40DKFZp686K191, STARING
Gene Symbols RNF40
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human RNF40. The exact sequence is proprietary.
Molecular Weight of Antigen 114 kDa
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Chromatin Research, Vision, Zinc Finger
Primary or Secondary Primary
Gene ID (Entrez) 9810
Test Specificity Human. Species reactivity based on sequence identity: Mouse (84%), Monkey (97%), Rat (84%), Bovine (87%).
Target Species Human
Content And Storage Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.