Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF43 Antibody (077), DyLight 650, Novus Biologicals™
SDP

Catalog No. NB072269 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB072269 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB072269 Supplier Novus Biologicals Supplier No. NBP290325C
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

RNF43 Monoclonal specifically detects RNF43 in Human samples. It is validated for ELISA.

Specifications

Antigen RNF43
Applications ELISA
Classification Monoclonal
Clone 77
Conjugate DyLight 650
Dilution ELISA
Formulation 50mM Sodium Borate
Gene Alias DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC
Host Species Rabbit
Immunogen This antibody was obtained from a rabbit immunized with purified, recombinant Human RNF43 (NP_060233.3; Met1-Tyr197).
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54894
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.