Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF43 Antibody (923227R), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. NB311074 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB311074 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB311074 Supplier Novus Biologicals Supplier No. FAB7964PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RNF43 Monoclonal antibody specifically detects RNF43 in Human samples. It is validated for Flow Cytometry

Specifications

Antigen RNF43
Applications Flow Cytometry
Classification Monoclonal
Clone 923227R
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS
Gene Alias DKFZp781H02126, DKFZp781H0392, FLJ20315, FLJ77466, FLJ99338, MGC125630, ring finger protein 43, RNF124, URCC
Host Species Mouse
Immunogen Human embryonic kidney cell line HEK293-derived recombinant human RNF43, Glu43-Val199, Accession # Q68DV7
Purification Method Protein A or G purified from cell culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer
Primary or Secondary Primary
Gene ID (Entrez) 54894
Target Species Human
Content And Storage Store at 4C in the dark. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.