Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF5 Antibody, Allophycocyanin, Novus Biologicals™
SDP

Catalog No. NB290731 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB290731 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB290731 Supplier Novus Biologicals Supplier No. NBP204014APC
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RNF5 Polyclonal antibody specifically detects RNF5 in Human samples. It is validated for Western Blot, Immunoprecipitation

Specifications

Antigen RNF5
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Conjugate APC
Dilution Western Blot, Immunoprecipitation
Formulation PBS
Gene Alias EC 6.3.2.-, G16, HsRma1, NG2, Protein G16, ring finger protein 5Ram1 homolog, RING5hsRma1, RMA1E3 ubiquitin-protein ligase RNF5
Host Species Rabbit
Immunogen Maps to a region between residue 1 and 50 of human Ring Finger Protein 5 using the numbering given in entry NP_008844.1 (GeneID 6048).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Biology, Immunology
Primary or Secondary Primary
Gene ID (Entrez) 6048
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.