Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RNF7 Antibody, Novus Biologicals™
SDP

Catalog No. NBP146159 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP146159 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP146159 Supplier Novus Biologicals Supplier No. NBP146159
Add to Cart
Add to Cart

Goat Polyclonal Antibody

RNF7 Polyclonal specifically detects RNF7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Peptide ELISA.

Specifications

Antigen RNF7
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), ELISA
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1 - 3 ug/mL, Immunohistochemistry 5 ug/mL, Immunohistochemistry-Paraffin 5 ug/mL, Peptide ELISA Detection limit 1:8000
Formulation Tris saline (20 mM Tris pH 7.3, 150 mM NaCl), 0.5% BSA with 0.02% Sodium Azide
Gene Accession No. Q9UBF6
Gene Alias CKBBP1zinc RING finger protein SAG, CKII beta-binding protein 1, Rbx2, Regulator of cullins 2, ring finger protein 7rbx2, ROC2RING-box protein 2, SAGsensitive to apoptosis, zinc RING finger protein SAG, regulator of cullins 2, Sensitive to apoptosis gene protein
Gene Symbols RNF7
Host Species Goat
Immunogen Peptide with sequence DACLRCQAENKQE corresponding to internal region according to NP_055060.1.
Molecular Weight of Antigen 13 kDa
Purification Method Affinity Purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9616
Test Specificity This antibody is expected to recognize isoform 1 (NP_055060.1).
Target Species Human
Content And Storage Store at -20C. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.